Assay Details
| Gene Symbol: FACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179931.4 | NP_850262.2 | 6 - 7 | 944 | 92 | |
| GenBank mRNA | AJ534971.1 | - | 6 - 7 | 742 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FACE2| Gene Symbol: FACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179931.4 | NP_850262.2 | 6 - 7 | 944 | 92 | |
| GenBank mRNA | AJ534971.1 | - | 6 - 7 | 742 | 92 | |
Set Membership: |
|