Assay Details
| Gene Symbol: TKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202764.1 | NP_001189693.1 | 11 - 12 | 2131 | 84 | |
| NM_001336615.1 | NP_001318368.1 | 10 - 11 | 2010 | 84 | ||
| NM_179942.4 | NP_850273.1 | 11 - 12 | 2279 | 84 | ||
| GenBank mRNA | AY064068.1 | - | 11 - 12 | 2074 | 84 | |
| AY096376.1 | - | 10 - 11 | 1927 | 84 | ||
More Information
Set Membership: |
|

TKI1
Genomic Map