Assay Details
| Gene Symbol: TKI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202764.1 | NP_001189693.1 | 9 - 10 | 1212 | 122 | |
| NM_001336615.1 | NP_001318368.1 | 8 - 9 | 1091 | 122 | ||
| NM_179942.4 | NP_850273.1 | 9 - 10 | 1360 | 122 | ||
| GenBank mRNA | AY064068.1 | - | 9 - 10 | 1155 | 122 | |
| AY096376.1 | - | 8 - 9 | 1008 | 122 | ||
More Information
Set Membership: |
|

TKI1
Genomic Map