Assay Details
| Gene Symbol: TRFL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036420.2 | NP_001031497.1 | 6 - 7 | 1608 | 129 | |
| NM_001336621.1 | NP_001324498.1 | 7 - 8 | 906 | 129 | ||
| NM_179944.3 | NP_850275.1 | 7 - 8 | 984 | 129 | ||
More Information
Set Membership: |
|

TRFL8
Genomic Map