Assay Details
| Gene Symbol: LCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036429.2 | NP_001031506.1 | 3 - 4 | 869 | 68 | |
| NM_001336677.1 | NP_001324059.1 | 2 - 3 | 1024 | 68 | ||
| NM_179956.2 | NP_850287.1 | 3 - 4 | 775 | 68 | ||
| NM_179957.3 | NP_850288.2 | 2 - 3 | 982 | 68 | ||
| GenBank mRNA | AK316808.1 | - | 3 - 4 | 766 | 68 | |
| AY081345.1 | - | 3 - 4 | 765 | 68 | ||
| BT002153.1 | - | 2 - 3 | 660 | 68 | ||
More Information
Set Membership: |
|

LCD1
Genomic Map