Assay Details
| Gene Symbol: LCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036429.2 | NP_001031506.1 | 4 - 5 | 1099 | 76 | |
| NM_001336677.1 | NP_001324059.1 | 3 - 4 | 1254 | 76 | ||
| NM_179956.2 | NP_850287.1 | 4 - 5 | 1005 | 76 | ||
| NM_179957.3 | NP_850288.2 | 3 - 4 | 1212 | 76 | ||
| GenBank mRNA | AK316808.1 | - | 4 - 5 | 996 | 76 | |
| AY081345.1 | - | 4 - 5 | 995 | 76 | ||
| BT002153.1 | - | 3 - 4 | 890 | 76 | ||
More Information
Set Membership: |
|

LCD1
Genomic Map