Assay Details
| Gene Symbol: LCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036429.2 | NP_001031506.1 | 7 - 8 | 1313 | 101 | |
| NM_001336677.1 | NP_001324059.1 | 6 - 7 | 1468 | 101 | ||
| NM_179956.2 | NP_850287.1 | 6 - 7 | 1313 | 101 | ||
| NM_179957.3 | NP_850288.2 | 5 - 6 | 1520 | 101 | ||
| GenBank mRNA | AK316808.1 | - | 7 - 8 | 1210 | 101 | |
| AY081345.1 | - | 6 - 7 | 1303 | 101 | ||
More Information
Set Membership: |
|

LCD1
Genomic Map