Assay Details
| Gene Symbol: AT2G38025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179959.4 | NP_850290.1 | 2 - 3 | 307 | 138 | |
| GenBank mRNA | AK117356.1 | - | 2 - 3 | 223 | 138 | |
| BT005253.1 | - | 2 - 3 | 192 | 138 | ||
| JQ013444.1 | - | 2 - 3 | 192 | 138 | ||
More Information
Set Membership: |
|

AT2G38025
Genomic Map