Assay Details
| Gene Symbol: AT2G38025 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336685.1 | NP_001325264.1 | 5 - 6 | 644 | 96 | |
| NM_179959.4 | NP_850290.1 | 6 - 7 | 612 | 96 | ||
| GenBank mRNA | AK117356.1 | - | 6 - 7 | 528 | 96 | |
| BT005253.1 | - | 6 - 7 | 497 | 96 | ||
| JQ013444.1 | - | 6 - 7 | 497 | 96 | ||
More Information
Set Membership: |
|

AT2G38025
Genomic Map