Assay Details
| Gene Symbol: CAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129360.4 | NP_565880.1 | 4 - 5 | 904 | 98 | |
| NM_179960.1 | NP_850291.1 | 4 - 5 | 818 | 98 | ||
| GenBank mRNA | AF056969.1 | - | 4 - 5 | 774 | 98 | |
More Information
Set Membership: |
|

CAC3
Genomic Map