Assay Details
| Gene Symbol: FAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129384.3 | NP_565886.1 | 12 - 13 | 1963 | 79 | |
| NM_179963.3 | NP_850294.1 | 12 - 13 | 1963 | 79 | ||
| GenBank mRNA | AY056301.1 | - | 12 - 13 | 1867 | 79 | |
| AY133852.1 | - | 12 - 13 | 1784 | 79 | ||
More Information
Set Membership: |
|

FAC1
Genomic Map