Assay Details
| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036436.1 | NP_001031513.1 | 13 - 14 | 1122 | 73 | |
| NM_179978.3 | NP_850309.1 | 13 - 14 | 1212 | 73 | ||
| GenBank mRNA | AY072346.1 | - | 13 - 14 | 1122 | 73 | |
| BT002547.1 | - | 13 - 14 | 1055 | 73 | ||
More Information
Set Membership: |
|

APC7
Genomic Map