Assay Details
| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179978.3 | NP_850309.1 | 16 - 17 | 1683 | 102 | |
| GenBank mRNA | AY072346.1 | - | 16 - 17 | 1593 | 102 | |
| BT002547.1 | - | 16 - 17 | 1526 | 102 | ||
More Information
Set Membership: |
|

APC7
Genomic Map