Assay Details
| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036436.1 | NP_001031513.1 | 4 - 5 | 395 | 152 | |
| NM_179978.3 | NP_850309.1 | 4 - 5 | 485 | 152 | ||
| GenBank mRNA | AY072346.1 | - | 4 - 5 | 395 | 152 | |
| BT002547.1 | - | 4 - 5 | 328 | 152 | ||
More Information
Set Membership: |
|

APC7
Genomic Map