Assay Details
| Gene Symbol: APC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036436.1 | NP_001031513.1 | 5 - 6 | 448 | 96 | |
| NM_179978.3 | NP_850309.1 | 5 - 6 | 538 | 96 | ||
| GenBank mRNA | AY072346.1 | - | 5 - 6 | 448 | 96 | |
| BT002547.1 | - | 5 - 6 | 381 | 96 | ||
More Information
Set Membership: |
|

APC7
Genomic Map