Assay Details
| Gene Symbol: AtCaN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336837.1 | NP_001318389.1 | 1 - 2 | 275 | 94 | |
| NM_201920.3 | NP_973649.1 | 1 - 2 | 313 | 94 | ||
More Information
Set Membership: |
|

AtCaN2
Genomic Map