Assay Details
| Gene Symbol: AtCaN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336837.1 | NP_001318389.1 | 4 - 5 | 554 | 68 | |
| NM_201920.3 | NP_973649.1 | 4 - 5 | 592 | 68 | ||
More Information
Set Membership: |
|

AtCaN2
Genomic Map