Assay Details
| Gene Symbol: AT1G01760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331304.1 | NP_001322029.1 | 4 - 5 | 850 | 69 | |
| NM_001331305.1 | NP_001322030.1 | 4 - 5 | 850 | 69 | ||
| NM_001331306.1 | NP_001322031.1 | 4 - 5 | 699 | 69 | ||
| NM_100059.3 | NP_171681.3 | 4 - 5 | 699 | 69 | ||
More Information
Set Membership: |
|

AT1G01760
Genomic Map