Assay Details
| Gene Symbol: PFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331308.1 | NP_001322242.1 | 4 - 5 | 861 | 96 | |
| NM_001331309.1 | NP_001322241.1 | 4 - 5 | 838 | 96 | ||
| NM_001331310.1 | NP_001322240.1 | 5 - 6 | 751 | 96 | ||
| NM_100068.3 | NP_171690.1 | 5 - 6 | 751 | 96 | ||
| GenBank mRNA | AF051326.1 | - | 5 - 6 | 716 | 96 | |
| AK117412.1 | - | 5 - 6 | 723 | 96 | ||
| BT005097.1 | - | 5 - 6 | 716 | 96 | ||
More Information
Set Membership: |
|

PFC1
Genomic Map