Assay Details
| Gene Symbol: AT1G01880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197958.1 | NP_001184887.1 | 1 - 2 | 317 | 93 | |
| NM_001331312.1 | NP_001320987.1 | 3 - 4 | 442 | 93 | ||
| NM_100069.3 | NP_171691.2 | 1 - 2 | 428 | 93 | ||
More Information
Set Membership: |
|

AT1G01880
Genomic Map