Assay Details
| Gene Symbol: SEC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197959.1 | NP_001184888.1 | 15 - 16 | 1424 | 92 | |
| NM_001331318.1 | NP_001320381.1 | 15 - 16 | 1593 | 92 | ||
| NM_001331319.1 | NP_001320383.1 | 14 - 15 | 1664 | 92 | ||
| NM_001331320.1 | NP_001320382.1 | 14 - 15 | 1658 | 92 | ||
| NM_001331321.1 | NP_001320384.1 | 9 - 10 | 952 | 92 | ||
| NM_100081.5 | NP_563643.4 | 15 - 16 | 1785 | 92 | ||
More Information
Set Membership: |
|

SEC1A
Genomic Map