Assay Details
| Gene Symbol: SEC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197959.1 | NP_001184888.1 | 17 - 18 | 1619 | 64 | |
| NM_001331318.1 | NP_001320381.1 | 17 - 18 | 1788 | 64 | ||
| NM_001331319.1 | NP_001320383.1 | 16 - 17 | 1859 | 64 | ||
| NM_001331320.1 | NP_001320382.1 | 16 - 17 | 1853 | 64 | ||
| NM_001331321.1 | NP_001320384.1 | 11 - 12 | 1147 | 64 | ||
| NM_100081.5 | NP_563643.4 | 17 - 18 | 1980 | 64 | ||
More Information
Set Membership: |
|

SEC1A
Genomic Map