Assay Details
| Gene Symbol: SEC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197959.1 | NP_001184888.1 | 5 - 6 | 424 | 101 | |
| NM_001331318.1 | NP_001320381.1 | 5 - 6 | 593 | 101 | ||
| NM_001331319.1 | NP_001320383.1 | 5 - 6 | 574 | 101 | ||
| NM_001331320.1 | NP_001320382.1 | 4 - 5 | 658 | 101 | ||
| NM_100081.5 | NP_563643.4 | 5 - 6 | 785 | 101 | ||
More Information
Set Membership: |
|

SEC1A
Genomic Map