Assay Details
| Gene Symbol: SEC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197959.1 | NP_001184888.1 | 9 - 10 | 849 | 92 | |
| NM_001331318.1 | NP_001320381.1 | 9 - 10 | 1018 | 92 | ||
| NM_001331319.1 | NP_001320383.1 | 8 - 9 | 1089 | 92 | ||
| NM_001331320.1 | NP_001320382.1 | 8 - 9 | 1083 | 92 | ||
| NM_001331321.1 | NP_001320384.1 | 3 - 4 | 377 | 92 | ||
| NM_100081.5 | NP_563643.4 | 9 - 10 | 1210 | 92 | ||
More Information
Set Membership: |
|

SEC1A
Genomic Map