Assay Details
| Gene Symbol: MAGO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100094.3 | NP_171716.1 | 1 - 2 | 269 | 66 | |
| GenBank mRNA | AY065157.1 | - | 1 - 2 | 252 | 66 | |
| AY087464.1 | - | 1 - 2 | 252 | 66 | ||
| AY114602.1 | - | 1 - 2 | 214 | 66 | ||
More Information
Set Membership: |
|

MAGO
Genomic Map