Assay Details
| Gene Symbol: GA2OX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331335.1 | NP_001323245.1 | 1 - 2 | 567 | 95 | |
| NM_100121.5 | NP_171742.1 | 1 - 2 | 568 | 95 | ||
| GenBank mRNA | AK228830.1 | - | 1 - 2 | 467 | 95 | |
| AY859741.1 | - | 1 - 2 | 404 | 95 | ||
| BT014886.1 | - | 1 - 2 | 404 | 95 | ||
More Information
Set Membership: |
|

GA2OX6
Genomic Map