Assay Details
| Gene Symbol: CLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100137.4 | NP_563657.1 | 2 - 3 | 505 | 75 | |
| GenBank mRNA | AB022326.1 | - | 2 - 3 | 298 | 75 | |
| AJ012278.1 | - | 2 - 3 | 268 | 75 | ||
| AY084394.1 | - | 2 - 3 | 346 | 75 | ||
| BT024859.1 | - | 2 - 3 | 245 | 75 | ||
More Information
Set Membership: |
|

CLPP5
Genomic Map