Assay Details
| Gene Symbol: CLPP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100137.4 | NP_563657.1 | 8 - 9 | 963 | 58 | |
| GenBank mRNA | AB022326.1 | - | 8 - 9 | 756 | 58 | |
| AJ012278.1 | - | 8 - 9 | 726 | 58 | ||
| AY084394.1 | - | 8 - 9 | 804 | 58 | ||
| BT024859.1 | - | 8 - 9 | 703 | 58 | ||
More Information
Set Membership: |
|

CLPP5
Genomic Map