Assay Details
| Gene Symbol: SPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331383.1 | NP_001322130.1 | 1 - 2 | 453 | 147 | |
| NM_100188.1 | NP_171805.1 | 1 - 2 | 187 | 147 | ||
More Information
Set Membership: |
|

SPI
Genomic Map