Assay Details
| Gene Symbol: SPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331381.1 | NP_001322129.1 | 8 - 9 | 1610 | 90 | |
| NM_001331382.1 | NP_001322128.1 | 8 - 9 | 1623 | 90 | ||
| NM_001331383.1 | NP_001322130.1 | 8 - 9 | 1230 | 90 | ||
| NM_100188.1 | NP_171805.1 | 8 - 9 | 964 | 90 | ||
More Information
Set Membership: |
|

SPI
Genomic Map