Assay Details
| Gene Symbol: AT1G03260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100208.4 | NP_171825.3 | 8 - 9 | 1032 | 89 | |
| GenBank mRNA | BT011222.1 | - | 8 - 9 | 720 | 89 | |
| BT011664.1 | - | 8 - 9 | 643 | 89 | ||
More Information
Set Membership: |
|

AT1G03260
Genomic Map