Assay Details
| Gene Symbol: BSU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331412.1 | NP_001321215.1 | 17 - 18 | 1890 | 97 | |
| NM_100227.6 | NP_171844.6 | 17 - 18 | 1890 | 97 | ||
| GenBank mRNA | AY372269.1 | - | 17 - 18 | 1890 | 97 | |
More Information
Set Membership: |
|

BSU1
Genomic Map