Assay Details
| Gene Symbol: LIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100230.7 | NP_171847.4 | 3 - 4 | 829 | 125 | |
| GenBank mRNA | AB044394.1 | - | 3 - 4 | 696 | 125 | |
| AJ420796.1 | - | 3 - 4 | 696 | 125 | ||
More Information
Set Membership: |
|

LIN2
Genomic Map