Assay Details
| Gene Symbol: SCAMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331416.1 | NP_001320329.1 | 4 - 5 | 359 | 64 | |
| NM_001331417.1 | NP_001320328.1 | 5 - 6 | 359 | 64 | ||
| NM_001331418.1 | NP_001320327.1 | 7 - 8 | 536 | 64 | ||
| NM_001331419.1 | NP_001320326.1 | 7 - 8 | 536 | 64 | ||
| NM_001331420.1 | NP_001320325.1 | 7 - 8 | 536 | 64 | ||
| NM_001331421.1 | NP_001320324.1 | 7 - 8 | 536 | 64 | ||
| NM_100237.1 | NP_563686.1 | 7 - 8 | 714 | 64 | ||
| GenBank mRNA | AY086966.1 | - | 7 - 8 | 714 | 64 | |
| BT024511.1 | - | 7 - 8 | 536 | 64 | ||
More Information
Set Membership: |
|

SCAMP4
Genomic Map