Assay Details
| Gene Symbol: AT1G03700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100249.5 | NP_563689.1 | 1 - 2 | 240 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G03700| Gene Symbol: AT1G03700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100249.5 | NP_563689.1 | 1 - 2 | 240 | 79 | |
Set Membership: |
|