Assay Details
| Gene Symbol: AT1G03760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100255.3 | NP_171872.2 | 4 - 5 | 861 | 87 | |
| GenBank mRNA | AK118726.1 | - | 4 - 5 | 861 | 87 | |
| BT005634.1 | - | 4 - 5 | 804 | 87 | ||
More Information
Set Membership: |
|

AT1G03760
Genomic Map