Assay Details
| Gene Symbol: AT1G03760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100255.3 | NP_171872.2 | 8 - 9 | 1105 | 60 | |
| GenBank mRNA | AK118726.1 | - | 8 - 9 | 1105 | 60 | |
| BT005634.1 | - | 8 - 9 | 1048 | 60 | ||
More Information
Set Membership: |
|

AT1G03760
Genomic Map