Assay Details
| Gene Symbol: AT1G03910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100272.4 | NP_171887.1 | 7 - 8 | 915 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G03910| Gene Symbol: AT1G03910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100272.4 | NP_171887.1 | 7 - 8 | 915 | 67 | |
Set Membership: |
|