Assay Details
| Gene Symbol: AT1G03910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197975.1 | NP_001184904.1 | 9 - 10 | 1108 | 63 | |
| NM_100272.4 | NP_171887.1 | 8 - 9 | 1015 | 63 | ||
More Information
Set Membership: |
|

AT1G03910
Genomic Map