Assay Details
| Gene Symbol: EIF2 GAMMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331466.1 | NP_001320247.1 | 5 - 6 | 818 | 94 | |
| NM_100298.3 | NP_171913.1 | 5 - 6 | 652 | 94 | ||
| GenBank mRNA | AF353097.1 | - | 5 - 6 | 497 | 94 | |
| AY092973.1 | - | 5 - 6 | 587 | 94 | ||
| BT002095.1 | - | 4 - 5 | 389 | 94 | ||
More Information
Set Membership: |
|

EIF2 GAMMA
Genomic Map