Assay Details
| Gene Symbol: EIF2 GAMMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331466.1 | NP_001320247.1 | 6 - 7 | 1047 | 62 | |
| NM_100298.3 | NP_171913.1 | 6 - 7 | 881 | 62 | ||
| GenBank mRNA | AF353097.1 | - | 6 - 7 | 726 | 62 | |
| AY092973.1 | - | 6 - 7 | 816 | 62 | ||
| BT002095.1 | - | 5 - 6 | 618 | 62 | ||
More Information
Set Membership: |
|

EIF2 GAMMA
Genomic Map