Assay Details
| Gene Symbol: EIF2 GAMMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331466.1 | NP_001320247.1 | 8 - 9 | 1524 | 89 | |
| NM_100298.3 | NP_171913.1 | 8 - 9 | 1358 | 89 | ||
| GenBank mRNA | AF353097.1 | - | 8 - 9 | 1203 | 89 | |
| AY092973.1 | - | 8 - 9 | 1293 | 89 | ||
| BT002095.1 | - | 7 - 8 | 1095 | 89 | ||
More Information
Set Membership: |
|

EIF2 GAMMA
Genomic Map