Assay Details
| Gene Symbol: TPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100300.5 | NP_171915.1 | 3 - 4 | 471 | 69 | |
| GenBank mRNA | AY080790.1 | - | 3 - 4 | 454 | 69 | |
| AY114043.1 | - | 3 - 4 | 303 | 69 | ||
More Information
Set Membership: |
|

TPR3
Genomic Map