Assay Details
| Gene Symbol: TPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100300.5 | NP_171915.1 | 6 - 7 | 940 | 72 | |
| GenBank mRNA | AY080790.1 | - | 6 - 7 | 923 | 72 | |
| AY114043.1 | - | 6 - 7 | 772 | 72 | ||
More Information
Set Membership: |
|

TPR3
Genomic Map