Assay Details
| Gene Symbol: AXR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100306.4 | NP_171921.1 | 3 - 4 | 892 | 74 | |
| GenBank mRNA | AF336916.1 | - | 3 - 4 | 588 | 74 | |
| AY070094.1 | - | 3 - 4 | 670 | 74 | ||
| AY117183.1 | - | 3 - 4 | 588 | 74 | ||
| U49073.1 | - | 3 - 4 | 649 | 74 | ||
More Information
Set Membership: |
|

AXR3
Genomic Map