Assay Details
| Gene Symbol: AT1G04420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100322.5 | NP_171937.1 | 1 - 2 | 341 | 104 | |
| GenBank mRNA | AY065361.1 | - | 1 - 2 | 247 | 104 | |
| AY122940.1 | - | 1 - 2 | 226 | 104 | ||
More Information
Set Membership: |
|

AT1G04420
Genomic Map