Assay Details
| Gene Symbol: AT1G04420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100322.5 | NP_171937.1 | 3 - 4 | 519 | 105 | |
| GenBank mRNA | AY065361.1 | - | 3 - 4 | 425 | 105 | |
| AY122940.1 | - | 3 - 4 | 404 | 105 | ||
More Information
Set Membership: |
|

AT1G04420
Genomic Map