Assay Details
| Gene Symbol: AT1G04420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100322.5 | NP_171937.1 | 5 - 6 | 800 | 88 | |
| GenBank mRNA | AY065361.1 | - | 5 - 6 | 706 | 88 | |
| AY122940.1 | - | 5 - 6 | 685 | 88 | ||
More Information
Set Membership: |
|

AT1G04420
Genomic Map