Assay Details
| Gene Symbol: AT1G04420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100322.5 | NP_171937.1 | 7 - 8 | 1111 | 87 | |
| GenBank mRNA | AY065361.1 | - | 7 - 8 | 1017 | 87 | |
| AY122940.1 | - | 7 - 8 | 996 | 87 | ||
More Information
Set Membership: |
|

AT1G04420
Genomic Map