Assay Details
| Gene Symbol: MAC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035881.1 | NP_001030958.1 | 10 - 11 | 1028 | 73 | |
| NM_100330.3 | NP_563708.1 | 10 - 11 | 1120 | 73 | ||
| GenBank mRNA | AY039940.1 | - | 10 - 11 | 1015 | 73 | |
| AY142564.1 | - | 10 - 11 | 854 | 73 | ||
More Information
Set Membership: |
|

MAC3A
Genomic Map